
N-Methyl-N-nitroso-p-toluenesulfonamide

Synonym(s):
p-Tolylsulfomethylnitrosamide, Diazald®, Diazomethane precursor, N-Methyl-N-(p-tolylsulfonyl)nitrosamide, N-Nitroso-p-toluenesulfomethylamide, N-Nitroso-N-methyl-p-toluenesulfonamide
Empirical Formula (Hill Notation):
C8H10N2O3S
Products
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Reaction Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing