
N-Boc-2-pyrroleboronic acid

Synonym(s):
N-Boc-2-pyrryl boronic acid
Empirical Formula (Hill Notation):
C9H14BNO4
CAS Number:
Molecular Weight:
211.02
Products
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing