Methyldopa
Synonym(s):
Methyl-DOPA sesquihydrate, 3-(3,4-Dihydroxyphenyl)-2-methyl-L-alanine, MK-351, Methyl-L-DOPA
Empirical Formula (Hill Notation):
C10H13NO4 · 1.5H2O
CAS Number:
Molecular Weight:
238.24
Products
Brand
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-3 of 3