
Meptyldinocap

Synonym(s):
(RS)-2-(1-Methylheptyl)-4,6-dinitrophenyl crotonate, 2-(1-Methylheptyl)-4,6-dinitrophenyl (2E)-2-butenoate
Empirical Formula (Hill Notation):
C18H24N2O6
CAS Number:
Molecular Weight:
364.39
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2