
Fumonisin B1

Synonym(s):
Fumonisin B1 from Fusarium moniliforme, Macrofusine
Empirical Formula (Hill Notation):
C34H59NO15
CAS Number:
Molecular Weight:
721.83
Products
Product Category
Brand
Formula Weight
Manufacturer
Format
Quality Segment
Application
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing