
Flamprop-M-isopropyl

Empirical Formula (Hill Notation):
C19H19ClFNO3
CAS Number:
Molecular Weight:
363.81
Products
Brand
Formula Weight
Manufacturer
Product Line
Quality Segment
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing