
Econazole nitrate salt

Synonym(s):
1-(2-[(4-Chlorophenyl)methoxy]-2-[2,4-dichlorophenyl]ethyl)-1H-imidazole
Empirical Formula (Hill Notation):
C18H15Cl3N2O · HNO3
Products
Product Category
Brand
Color
Formula Weight
Physical Form
Activity Spectrum
Quality Segment
Mode of Action
Showing 1-1 of 1
Compare
Product Number
Description
Pricing