
D-Pinitol

Synonym(s):
3-O-Methyl-D-chiro-inositol
Empirical Formula (Hill Notation):
C7H14O6
CAS Number:
Molecular Weight:
194.18
Products
Product Category
Formula Weight
Melting Point (°C)
Physical Form
Purity
Format
Quality Segment
Functional Group
Markush Class
Markush Group
Optical Isomer
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing