
Diphenhydramine Impurity B

Synonym(s):
2-[(RS)-(4-Methylphenyl)phenylmethoxy]-N,N-dimethylethanamine hydrochloride, 2-[(4-Methyl-α-phenylbenzyl)oxy]ethyl(dimethyl)ammonium chloride, 4-Methyldiphenhydramine hydrochloride, N,N-Dimethyl-2-[(4-methylphenyl)phenylmethoxy]ethanamin hydrochloride, Neobenodine
Empirical Formula (Hill Notation):
C18H23NO · HCl
CAS Number:
Molecular Weight:
305.84
Products
Brand
Formula Weight
Physical Form
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing