
Benzil

Synonym(s):
Benzil, Dibenzoyl, Diphenylethanedione
Linear Formula:
C6H5COCOC6H5
CAS Number:
Molecular Weight:
210.23
Products
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Quality Segment
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing