Skip to Content
Merck
Almotriptan Related Compound A certified reference material, pharmaceutical secondary standard

Almotriptan Related Compound A

Almotriptan Related Compound A certified reference material, pharmaceutical secondary standard
Synonym(s):
{3-[2-(Dimethylamino)ethyl]-5-[(pyrrolidine-1-yl)sulfonylmethyl]-1H-indol-1-yl}methanol
Empirical Formula (Hill Notation):
C18H27N3O3S
CAS Number:
Molecular Weight:
365.49

Products

Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade