Skip to Content
Merck
All Photos(1)

Key Documents

580222

Sigma-Aldrich

Tau Aggregation Inhibitor, ATPZ, Cpd16

Synonym(s):

Tau Aggregation Inhibitor, ATPZ, Cpd16, 5-amino-3-(4-chlorophenyl)-N-cyclopropyl-4-oxo-3,4-dihydrothieno[3,4-d]pyridazine-1-carboxamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H13ClN4O2S
Molecular Weight:
360.82
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥95% (HPLC)

Quality Level

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

orange

solubility

DMSO: 50 mg/mL

shipped in

wet ice

storage temp.

−20°C

General description

A water soluble (32 µM in water) BBB-permeable ATPZ (aminothienopyridazine) tau aggregation inhibitor that exhibited IC50 of 8.1 µM in a heparin-induced tau assembly assay (monitored by thioflavine-T binding and fluorescence), and was further validated in vitro to reduce tau fibril formation (tau fragment K18PL, and full length tau40) by >50% in sedimentation assays. It also displayed very good B/P ratio (1.6) in a mice full PK study with good oral bioavailability and good metabolic stability. No cytotoxicity was observed at up to 100 µM in HEK-293 cells.
A water soluble (32uM in water) BBB-permeable ATPZ (aminothienopyridazine) tau aggregation inhibitor that exhibited IC50 of 8.1uM in a heparin-induced tau assembly assay (monitored by thioflavine-T binding and fluorescence), and was further validated in vitro to reduce tau fibril formation (tau fragment K18PL, and full length tau40) by >50% in sedimentation assays. It also displayed very good B/P ratio (1.6) in a mice full PK study with good oral bioavailability and good metabolic stability. No cytotoxicity was observed at up to 100uM in HEK-293 cells.

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Ballatore, C., et al. 2010. J. Med. Chem.53, 3739.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service