- Occurrence and distribution of organophosphate esters in urban soils of the subtropical city, Guangzhou, China.
Occurrence and distribution of organophosphate esters in urban soils of the subtropical city, Guangzhou, China.
Chemosphere (2017-03-04)
Kunyan Cui, Jiaxin Wen, Feng Zeng, Shuocong Li, Xi Zhou, Zunxiang Zeng
PMID28254716
ZUSAMMENFASSUNG
67 soil samples from the road greenbelts, the paddy/vegetable fields, the parks, the commercial and residential areas in the subtropical city, Guangzhou, China, were collected and analyzed for 11 organophosphate esters (OPEs) and triphenylphosphine oxide (TPPO). OPEs were detected in all soil samples analyzed, which indicate that OPEs are ubiquitous environmental contaminants. The ∑
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
SKU
Packungsgröße
Verfügbarkeit
Preis
Menge