Merck
  • Loss of cytokeratin 20 and acquisition of thyroid transcription factor-1 expression in a merkel cell carcinoma metastasis to the brain.
Loss of cytokeratin 20 and acquisition of thyroid transcription factor-1 expression in a merkel cell carcinoma metastasis to the brain.
The American Journal of dermatopathology (2014-06-06)
Sara C Shalin, Christopher P Cifarelli, James Y Suen, Ling Gao
ZUSAMMENFASSUNG
Merkel cell carcinoma (MCC) is recognized by its morphologic features and by its classic immunophenotypic properties. Although MCCs demonstrating nonclassic immunoreactivities have been described, a case documenting a change in immunophenotype during the course of disease progression has not been previously reported. We report a case of MCC that initially demonstrated cytokeratin 20 positivity but lost expression in subsequent metastases. Likewise, thyroid transcription factor-1 was initially negative in the tumor but expression was present in metastatic lesions.