Merck
  • CLIF, a novel cycle-like factor, regulates the circadian oscillation of plasminogen activator inhibitor-1 gene expression.
CLIF, a novel cycle-like factor, regulates the circadian oscillation of plasminogen activator inhibitor-1 gene expression.
The Journal of biological chemistry (2000-10-06)
K Maemura, S M de la Monte, M T Chin, M D Layne, C M Hsieh, S F Yet, M A Perrella, M E Lee
ZUSAMMENFASSUNG
The onset of myocardial infarction occurs frequently in the early morning, and it may partly result from circadian variation of fibrinolytic activity. Plasminogen activator inhibitor-1 activity shows a circadian oscillation and may account for the morning onset of myocardial infarction. However, the molecular mechanisms regulating this circadian oscillation remain unknown. Recent evidence indicates that basic helix-loop-helix (bHLH)/PAS domain transcription factors play a crucial role in controlling the biological clock that controls circadian rhythm. We isolated a novel bHLH/PAS protein, cycle-like factor (CLIF) from human umbilical vein endothelial cells. CLIF shares high homology with Drosophila CYCLE, one of the essential transcriptional regulators of circadian rhythm. CLIF is expressed in endothelial cells and neurons in the brain, including the suprachiasmatic nucleus, the center of the circadian clock. In endothelial cells, CLIF forms a heterodimer with CLOCK and up-regulates the PAI-1 gene through E-box sites. Furthermore, Period2 and Cryptochrome1, whose expression show a circadian oscillation in peripheral tissues, inhibit the PAI-1 promoter activation by the CLOCK:CLIF heterodimer. These results suggest that CLIF regulates the circadian oscillation of PAI-1 gene expression in endothelial cells. In addition, the results potentially provide a molecular basis for the morning onset of myocardial infarction.