Merck
  • [Influence of fluoxetine and paroxetine on anxiety-like behavior in young and adult prenatally stressed male rats].
[Influence of fluoxetine and paroxetine on anxiety-like behavior in young and adult prenatally stressed male rats].
Patologicheskaia fiziologiia i eksperimental'naia terapiia (2014-03-20)
N E Ordyan, S G Pivina, Yu O Fedotova, V K Akulova
ZUSAMMENFASSUNG
The aim of the present work was an estimation of effects of chronic administration of selective serotonin reuptake inhibitors--fluoxetine (5.0 mg/kg, p.o.) and paroxetine (5.0 mg/kg, p.o.) for 14 days of postnatal period on anxiety-like behavior in the prenatally stressed male rats during pubertal period (1,5 month) and the adult state (3 month). Chronic paroxetine administration to females failed to change an anxiety-like behavior independently from age. On the contrary, administration of fluoxetine resulted in modulating influence on the anxiety-like behavior of prenatally stressed rats dependently from age: anxiolytic effect was noted in young males, while anxiogenic effect was observed in the adult male rats.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Paroxetin-hydrochlorid (wasserfrei), European Pharmacopoeia (EP) Reference Standard