Merck
  • Determination of 5-methoxyindoles in pineal gland and plasma samples by high-performance liquid chromatography with electrochemical detection.
Determination of 5-methoxyindoles in pineal gland and plasma samples by high-performance liquid chromatography with electrochemical detection.
Journal of chromatography (1991-03-08)
F Raynaud, P Pévet
ZUSAMMENFASSUNG
A liquid chromatographic analysis with electrochemical detection of 5-methoxytryptamine, 5-methoxytryptophol, 5-methoxyindoleacetic acid and melatonin is described. Optimal elution conditions were determined by studying several variables: pH, buffer salt, counter ion and organic modifier. Measurement of 5-methoxyindoles in the pineal gland and plasma of hamsters has been performed after extraction. This method is specific and sensitive, and enables detection of 5-methoxyindoles in a pool of two hamster pineal glands. This is also the first time that these three 5-methoxyindoles have been measured simultaneously in plasma.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
5-Methoxy-3-Indolessigsäure, 98%