Merck
  • Silver(I)-catalyzed addition of zirconocenes to glycal epoxides. A new synthesis of alpha-C-glycosides.
Silver(I)-catalyzed addition of zirconocenes to glycal epoxides. A new synthesis of alpha-C-glycosides.
Organic letters (2005-01-28)
Peter Wipf, Joshua G Pierce, Nian Zhuang
ZUSAMMENFASSUNG
[reaction: see text] Hydrozirconation of terminal alkynes, followed by AgClO4-catalyzed in situ addition of the resultant alkenylzirconocenes to 1,2-anhydrosugars (glycal epoxides) leads to alpha-C-glycosides in moderate to high yields.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Bis(cyclopentadienyl)zirconium(IV)-dichlorid, ≥98%
SKU
Packungsgröße
Verfügbarkeit
Preis
Menge