Merck
  • Methylmercury exposure induces ROS/Akt inactivation-triggered endoplasmic reticulum stress-regulated neuronal cell apoptosis.
Methylmercury exposure induces ROS/Akt inactivation-triggered endoplasmic reticulum stress-regulated neuronal cell apoptosis.
Toxicology (2019-07-23)
Yao-Pang Chung, Cheng-Chieh Yen, Feng-Cheng Tang, Kuan-I Lee, Shing-Hwa Liu, Chin-Ching Wu, Shang-Shu Hsieh, Chin-Chuan Su, Chun-Ying Kuo, Ya-Wen Chen
ZUSAMMENFASSUNG
Epidemiological studies have positively linked mercury exposure and neurodegenerative diseases (ND). Methylmercury (MeHg), an organic form of mercury, is a ubiquitous and potent environmental neurotoxicant that easily crosses the blood-brain barrier and causes irreversible injury to the central nervous system (CNS). However, the molecular mechanisms underlying MeHg-induced neurotoxicity remain unclear. Here, the present study found that Neuro-2a cells underwent apoptosis in response to MeHg (1-5 μM), which was accompanied by increased phosphatidylserine (PS) exposure on the outer cellular membrane leaflets, caspase-3 activity, and the activation of caspase cascades and poly (ADP-ribose) polymerase (PARP). Exposure of Neuro-2a cells to MeHg also triggered endoplasmic reticulum (ER) stress, which was identified via several key molecules (including: glucose-regulated protein (GRP)78, GRP94, C/EBP homologous protein (CHOP) X-box binding protein(XBP)-1, protein kinase R-like ER kinase (PERK), eukaryotic initiation factor 2α (eIF2α), inositol-requiring enzyme(IRE)-1, activation transcription factor(AFT)4, and ATF6. Transfection with GRP78-, GRP94-, CHOP-, and XBP-1-specific small interfering (si)RNA significantly suppressed the expression of these proteins, and attenuated cytotoxicity and caspase-12, -7, and -3 activation in MeHg-exposed cells. Furthermore, MeHg dramatically decreased Akt phosphorylation, and the overexpression of activation of Akt1 (myr-Akt1) could significantly prevent MeHg-induced Akt inactivation, as well as apoptotic and ER stress-related signals. Pretreatment with the antioxidant N-acetylcysteine (NAC) effectively prevented MeHg-induced neuronal cell reactive oxygen species (ROS) generation, apoptotic and ER stress-related signals, and Akt inactivation. Collectively, these results indicate that MeHg exerts its cytotoxicity in neurons by inducing ROS-mediated Akt inactivation up-regulated ER stress, which induces apoptosis and ultimately leads to cell death.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
N-Acetyl-Asp-Glu-Val-Asp-7-amido-4-methylcoumarin, ≥97% (HPLC), powder
SKU
Packungsgröße
Verfügbarkeit
Preis
Menge
Sigma-Aldrich
Amyloid Protein Non-Aβ Component, ≥80% (HPLC)
SKU
Packungsgröße
Verfügbarkeit
Preis
Menge