Merck
SYNF431125
Synthware three neck round bottom flask with angled side necks
25 mL, center joint: ST/NS 14/20, side joint: ST/NS 14/20
Synonym(e):
Synthware F431125, 3-neck round flask
About This Item
UNSPSC-Code:
41121804
NACRES:
NB.51
Materialien
borosilicate glass flask
round bottom
three-neck (angled side necks)
Verbindung
center joint: ST/NS14/20
side joint: ST/NS14/20
Verpackung
pkg of 1 ea
Hersteller/Markenname
Synthware F431125
Kapazität
25 mL
25 mL
Allgemeine Beschreibung
Round bottom with joints. Heavy wall design. All sizes are hand-blown from tubing to ensure uniform wall thickness.
Rechtliche Hinweise
Synthware is a trademark of Kemtech America Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)