Merck
Tec Kinases
Tec kinases represent the second largest family of nonreceptor tyrosine kinases and are activated in response to cellular stimulation by antigen receptors, integrins, growth factors, cytokines and G protein-coupled receptors. The mammalian Tec family consists of five members: Tec, Btk, Itk/Emt/Tsk, Rlk/Txk and Bmx/Etk. Tec kinases are defined by a common protein domain organization including a COOH-terminal kinase domain, preceded by Src homology-2 and 3 protein interaction domains and a Tec homology domain that includes one or two proline-rich regions that interact intramolecularly or intermolecularly with SH3 domains and contribute to kinase regulation. Importantly, most Tec kinases possess an amino terminal pleckstrin homology (PH) domain that distinguishes them from all other identified tyrosine kinases. The Tec kinases' PH domains bind to phosphatidylinositol (3,4,5) trisphosphate (PIP3), and are therefore regulated by PI3 kinase and the phosphatases SHIP and PTEN. The atypical Tec kinase Rlk/Txk lacks a PH domain and instead contains a palmitoylated series of cysteines.
With some exceptions, Tec kinases are expressed primarily in cells of hematopoietic lineages. Btk is expressed in most hematopoietic cells except T cells, whereas Itk expression is limited to mast cells, T-, NK-, and NKT cells, and Rlk is restricted to T- and mast cells. In contrast, Tec is most widely expressed and is found in liver, developing embryo, brain, endothelium and melanocytes, in addition to hematopoietic cells. Bmx is expressed in granulocytes, monocytes, and in cells of epithelial and endothelial lineages.
Activation of Tec kinases requires two major steps: 1) membrane targeting, via interactions of their PH domains with PIP3 or other proteins and 2) tyrosine phosphorylation within the kinase activation loop. Protein interactions via the SH2 and SH3 domains may also be required to disrupt intramolecular interactions and to localize the kinases in signaling complexes.
Although Tec kinases are activated by many receptors, their functions are best understood downstream of lymphocyte antigen receptors. Notably, mutations affecting Btk cause the human primary immunodeficiency, X-linked agammaglobulinemia, as well as the mouse mutant x-linked immunodeficiency, xid, characterized by impaired B cell development and function. Similarly, mutations disrupting Itk or Itk and Rlk in mice cause defective T lymphocyte development and function associated with reduced antigen receptor induced proliferation, cytokine production, adhesion and migration. Btk and Itk are required for the phosphorylation and full activation of PLC-γ and downstream readouts including mobilization of calcium and activation of MAP kinases and downstream transcription factors, including NFATs, AP-1 and NFkB. Btk also interacts with and is cross-regulated by PKC-β. Additional roles for Tec kinases in T cells include regulation of the actin cytoskeleton, adhesion and migration. Upon antigen receptor activation, Rlk, Itk and Btk can translocate to the nucleus, suggesting direct effects on transcription.
In T cells, mutation of the Tec kinases neither prevents T cell development nor signaling but instead alters the efficiency or type of T cells responses. In particular, Itk-deficiency impairs TH2 responses associated with allergy and asthma, making Itk an attractive therapeutic target for such diseases. Several inhibitors of Itk have been described; data from three highly selective Itk antagonists demonstrate putative therapeutic use for allergic-induced asthma.
In other cell types Bmx/Etk and Tec participate in the regulation of Rho and serum response factor in response to Gα12. Tec is activated in response to multiple cytokine and growth factor receptors and has also been linked to the actin cytoskeleton via interactions with Vav. Bmx/Etk also participates in signaling from integrins and roles in wound healing and cardioprotection were also recently described. Bmx Etk is also required for phosphorylation of STAT-3 in cellular transformation by Src, suggesting potential therapeutic uses for Tec kinase inhibitors in cancer.
The Table below contains accepted modulators and additional information. For a list of additional products, see the "Similar Products" section below.
Footnotes
a)
 Multiple isoforms exist due to alternate initiation start sites or splicing variants.
b) Isoform Tec IV noted here, Tec isoforms I-IV have been reported for both human and mouse.
Similar Products
Loading
References
1.
Andreotti AH, Schwartzberg PL, Joseph RE, Berg LJ. 2010. T-Cell Signaling Regulated by the Tec Family Kinase, Itk. Cold Spring Harbor Perspectives in Biology. 2(7):a002287-a002287. https://doi.org/10.1101/cshperspect.a002287
2.
August A, Ragin MJ. 2012. Regulation of T-cell Responses and Disease by Tec Kinase Itk. International Reviews of Immunology. 31(2):155-165. https://doi.org/10.3109/08830185.2012.668981
3.
Berg LJ, Finkelstein LD, Lucas JA, Schwartzberg PL. 2005. TEC FAMILY KINASES IN T LYMPHOCYTE DEVELOPMENT AND FUNCTION. Annu. Rev. Immunol.. 23(1):549-600. https://doi.org/10.1146/annurev.immunol.22.012703.104743
4.
Berg LJ. 2007. Signalling through TEC kinases regulates conventional versus innate CD8+ T-cell development. Nat Rev Immunol. 7(6):479-485. https://doi.org/10.1038/nri2091
5.
Felices M, Falk M, Kosaka Y, Berg LJ. 2007. Tec Kinases in T Cell and Mast Cell Signaling.145-184. https://doi.org/10.1016/s0065-2776(06)93004-1
6.
Finkelstein LD, Schwartzberg PL. 2004. Tec kinases: shaping T-cell activation through actin. Trends in Cell Biology. 14(8):443-451. https://doi.org/10.1016/j.tcb.2003.07.001
7.
Fluckiger A, Li Z, Kato RM, Wahl MI, Ochs HD, Longnecker R, Kinet J, Witte ON, Scharenberg AM, Rawlings DJ. 1998. Btk/Tec kinases regulate sustained increases in intracellular Ca2+ following B-cell receptor activation. EMBO J. 17(7):1973-1985. https://doi.org/10.1093/emboj/17.7.1973
8.
Garçon F, Nunès JA. Travel Informations on the TEC Kinases during Lymphocyte Activation.15-27. https://doi.org/10.1007/0-387-34132-3_2
9.
Koprulu AD, Ellmeier W. 2009. The Role of Tec Family Kinases in Mononuclear Phagocytes. Crit Rev Immunol. 29(4):317-333. https://doi.org/10.1615/critrevimmunol.v29.i4.30
10.
Lin T, McIntyre KW, Das J, Liu C, O'Day KD, Penhallow B, Hung C, Whitney GS, Shuster DJ, Yang X, et al. 2004. Selective Itk Inhibitors Block T-Cell Activation and Murine Lung Inflammation. Biochemistry. 43(34):11056-11062. https://doi.org/10.1021/bi049428r
11.
Mano H. 1999. Tec family of protein-tyrosine kinases: an overview of their structure and function. Cytokine & Growth Factor Reviews. 10(3-4):267-280. https://doi.org/10.1016/s1359-6101(99)00019-2
12.
Qiu Y, Kung H. 2000. Signaling network of the Btk family kinases. Oncogene. 19(49):5651-5661. https://doi.org/10.1038/sj.onc.1203958
13.
Scharenberg AM, Kinet J. 1998. PtdIns-3,4,5-P3. Cell. 94(1):5-8. https://doi.org/10.1016/s0092-8674(00)81214-3
14.
Schmidt U, Boucheron N, Unger B, Ellmeier W. 2004. The Role of Tec Family Kinases in Myeloid Cells. Int Arch Allergy Immunol. 134(1):65-78. https://doi.org/10.1159/000078339
15.
Smith CE, Islam TC, Mattsson PT, Mohamed AJ, Nore BF, Vihinen M. 2001. The Tec family of cytoplasmic tyrosine kinases: mammalian Btk, Bmx, Itk, Tec, Txk and homologs in other species. Bioessays. 23(5):436-446. https://doi.org/10.1002/bies.1062
16.
Takata M, Kurosaki T. 1996. A role for Bruton's tyrosine kinase in B cell antigen receptor-mediated activation of phospholipase C-gamma 2.. 184(1):31-40. https://doi.org/10.1084/jem.184.1.31
17.
Vassilev A, Uckun F. 2004. Therapeutic Potential of Inhibiting Brutons Tyrosine Kinase, (BTK). CPD. 10(15):1757-1766. https://doi.org/10.2174/1381612043384475
18.
Vihinen M. 2000. Bruton tyrosine kinase BTK in X-linked agammaglobulinemia XLA. Front Biosci. 5(3):d917-928. https://doi.org/10.2741/a560