Merck
  • Study on molecularly imprinted nanoparticle modified microplates for pseudo-ELISA assays.
Study on molecularly imprinted nanoparticle modified microplates for pseudo-ELISA assays.
Talanta (2017-11-16)
Lucia Cenci, Chiara Piotto, Paolo Bettotti, Alessandra Maria Bossi
ABSTRACT
Nanosized Molecularly Imprinted Polymers (nanoMIPs) are designed artificial nanoreceptors with a predetermined selectivity and specificity for a given analyte, lately proposed as a replacement to antibodies in immunoassays. The nanoMIP-plate preparation based on nanoparticle adsorption was studied with the aim to rationally identify and discuss the critical points in the nanoMIP-assay development, in an example based on the iron homeostasis biomarker hepcidin and hepcidin-specific nanoMIPs (K
Sigma-Aldrich
N-tert-Butylacrylamide, 97%