- Biredox ionic liquids: new opportunities toward high performance supercapacitors.
Biredox ionic liquids: new opportunities toward high performance supercapacitors.
Faraday discussions (2017-09-25)
C Bodin, E Mourad, D Zigah, S Le Vot, S A Freunberger, F Favier, O Fontaine
PMID28936498
ABSTRACT
Nowadays commercial supercapacitors are based on purely capacitive storage at the porous carbons that are used for the electrodes. However, the limits that capacitive storage imposes on energy density calls to investigate new materials to improve the capacitance of the device. This new type of electrodes (e.g., RuO
MATERIALS