Merck
Augmented gene expression triggered by Na
Cell calcium (2017-11-14)
Larisa V Smolyaninova, Svetlana V Koltsova, Svetlana V Sidorenko, Sergei N Orlov
ABSTRACT
In rat vascular smooth muscle cells (RVSMC), 3-h Na
Sigma-Aldrich
W-13, ≥98% (HPLC)