Merck
  • Melanin quantification by in vitro and in vivo analysis of near-infrared fluorescence.
Melanin quantification by in vitro and in vivo analysis of near-infrared fluorescence.
Pigment cell & melanoma research (2017-08-15)
Sunil Kalia, Jianhua Zhao, Haishan Zeng, David McLean, Nikiforos Kollias, Harvey Lui
ABSTRACT
Objective measurements of melanin can provide important information for differentiating melanoma from benign pigmented lesions and in assessing pigmentary diseases. Herein, we evaluate near-infrared (NIR) fluorescence as a possible tool to quantify melanin. Various concentrations of in vitro Sepia melanin in tissue phantoms were measured with NIR fluorescence and diffuse reflectance spectroscopy. Similar optic measurements were conducted in vivo on 161 normal human skin sites. Diffuse reflectance spectroscopy was used to quantify the melanin content via Stamatas-Kollias algorithm. At physiologic concentrations, increasing in vitro melanin concentrations demonstrated higher fluorescence that was linearly correlated (R
Sigma-Aldrich
Melanin from Sepia officinalis, 99% (TLC)