- Photodegradation of some brominated and phenolic micropollutants in raw hospital wastewater with CeO
Photodegradation of some brominated and phenolic micropollutants in raw hospital wastewater with CeO
Water science and technology : a journal of the International Association on Water Pollution Research (2017-11-24)
Delia Teresa Sponza, Gökçe Güney
PMID29168700
ABSTRACT
In this study, the degradations of 2,3,4,5,6-pentabromotoluene (PBT), 2,3,4,5,6-pentabromoethyl benzene (PBEB), triclosan (TCS) and gemfibrozil (GFZ) in raw hospital wastewater were investigated with cerium (IV) oxide and titanium (IV) oxide nanoparticles considering the mechanisms of adsorption, photolysis, and photocatalysis with UV-C lamps. The effects of nano-CeO