Merck
  • Observation of the Marcus Inverted Region of Electron Transfer from Asymmetric Chemical Doping of Pristine (n,m) Single-Walled Carbon Nanotubes.
Observation of the Marcus Inverted Region of Electron Transfer from Asymmetric Chemical Doping of Pristine (n,m) Single-Walled Carbon Nanotubes.
Journal of the American Chemical Society (2017-10-08)
Yuichiro Kunai, Albert Tianxiang Liu, Anton L Cottrill, Volodymyr B Koman, Pingwei Liu, Daichi Kozawa, Xun Gong, Michael S Strano
ABSTRACT
The concept of electrical energy generation based on asymmetric chemical doping of single-walled carbon nanotube (SWNT) papers is presented. We explore 27 small, organic, electron-acceptor molecules that are shown to tune the output open-circuit voltage (V
Sigma-Aldrich
cGMP Enzyme Immunoassay Kit, Direct, sufficient for 96 assays