Merck
  • Temporal Properties of Flicker ERGs in Rabbit Model of Retinitis Pigmentosa.
Temporal Properties of Flicker ERGs in Rabbit Model of Retinitis Pigmentosa.
Investigative ophthalmology & visual science (2017-10-28)
Satoshi Okado, Shinji Ueno, Taro Kominami, Ayami Nakanishi, Daiki Inooka, Akira Sayo, Mineo Kondo, Hiroko Terasaki
ABSTRACT
We determined the effects of a remodeled inner retina on the flicker electroretinograms (ERGs) in a rabbit eye at an advanced stage of inherited retinal degeneration. Six wild-type (WT) and four rhodopsin P347L transgenic (Tg) rabbits were studied at 18 months of age. Flicker ERGs were elicited by sinusoidal stimuli at frequencies of 3.906 to 50.781 Hz. To block the ON and OFF retinal pathways, 2-amino-4-phosphonobutyric acid (APB), and 6-cyano-7-nitroquinoxaline-2, 3(1H, 4H)-dione (CNQX), respectively, were injected intravitreally. The amplitudes and phases of the fundamental components of the pre- and postdrug ERGs were analyzed. The postsynaptic APB (ON-) and CNQX (OFF-) sensitive components were determined by examining the phases and amplitude vectors. The temporal properties of the Tg rabbits were different from those of the WT rabbits and had unique features; at 3.906 Hz, the amplitude was depressed but it increased by more than 3.5-fold at 15.625 Hz. The reduction of the amplitude at 3.906 Hz in Tg rabbits was caused by a cancelation of the ON and OFF components by a phase difference of 180°. On the other hand, an increase in the amplitude at 15.625 Hz in Tg rabbits was caused by the summation of the ON and OFF components, which had an approximate 120° phase difference. The temporal properties of the flicker ERGs of Tg rabbits were affected markedly by the remodeling of the retinal neurons. Evaluations of the flicker ERGs in RP eyes must be done with careful considerations of the current findings.
Sigma-Aldrich
CNQX, ≥98% (HPLC), solid
Sigma-Aldrich
(±)-2-Amino-4-phosphonobutyric acid, solid