- Probing GATA factor function in mouse Leydig cells via testicular injection of adenoviral vectors.
Probing GATA factor function in mouse Leydig cells via testicular injection of adenoviral vectors.
Reproduction (Cambridge, England) (2017-07-16)
Gervette M Penny, Rebecca B Cochran, Marjut Pihlajoki, Antti Kyrönlahti, Anja Schrade, Merja Häkkinen, Jorma Toppari, Markku Heikinheimo, David B Wilson
PMID28710293
ABSTRACT
Testicular Leydig cells produce androgens essential for proper male reproductive development and fertility. Here, we describe a new Leydig cell ablation model based on Cre/Lox recombination of mouse