Merck
  • Low expression of SerpinB2 is associated with reduced survival in lung adenocarcinomas.
Low expression of SerpinB2 is associated with reduced survival in lung adenocarcinomas.
Oncotarget (2017-12-07)
Maria Ramnefjell, Christina Aamelfot, Lars Helgeland, Lars A Akslen
ABSTRACT
Lung cancer is a leading cause of cancer deaths worldwide and new biomarkers are of utmost importance. Studies have indicated that the anti-plasminogen activators SerpinB2 and Neuroserpin, and the adhesion molecule L1CAM, have a coordinated impact on development of metastasis. Here, we examined whether expression of these markers was associated with clinico-pathologic characteristics and prognosis in resected non-small cell lung cancer (NSCLC). Surgical specimens from 438 NSCLC patients treated at Haukeland University Hospital, Bergen, Norway (1993-2010) were included (median age 68 years; 213 adenocarcinomas, 135 squamous cell carcinomas, 90 others). Representative tumor sections were stained for SerpinB2, Neuroserpin, and L1CAM. Low expression of SerpinB2 was associated with reduced lung cancer specific survival (LCSS) in adenocarcinomas (
Sigma-Aldrich
Anti-SERPINI1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution