Merck
Signals of vagal circuits engaging with AKT1 in α7 nAChR
Cell discovery (2017-05-23)
Caiqi Zhao, Xi Yang, Emily M Su, Yuanyuan Huang, Ling Li, Michael A Matthay, Xiao Su
ABSTRACT
Vagal circuits-α7 nAChR (α7 nicotinic acetylcholine receptor, coded by
Sigma-Aldrich
(−)-Nicotine hemisulfate salt, ≥95% (TLC), ~35 % (w/v) in H2O (based on free base)