Merck
  • SIRT3 protects hepatocytes from oxidative injury by enhancing ROS scavenging and mitochondrial integrity.
SIRT3 protects hepatocytes from oxidative injury by enhancing ROS scavenging and mitochondrial integrity.
Cell death & disease (2017-10-27)
Jingxin Liu, Dan Li, Tian Zhang, Qiang Tong, Richard Dequan Ye, Ligen Lin
ABSTRACT
Evidences of oxidative stress and mitochondrial dysfunction have been recognized in most of clinical and experimental liver diseases. SIRT3, a member of NAD
Sigma-Aldrich
MISSION® esiRNA, targeting human SIRT3