Merck
  • Chromatin-Associated Protein SIN3B Prevents Prostate Cancer Progression by Inducing Senescence.
Chromatin-Associated Protein SIN3B Prevents Prostate Cancer Progression by Inducing Senescence.
Cancer research (2017-08-16)
Anthony J Bainor, Fang-Ming Deng, Yu Wang, Peng Lee, David J Cantor, Susan K Logan, Gregory David
ABSTRACT
Distinguishing between indolent and aggressive prostate adenocarcinoma remains a priority to accurately identify patients who need therapeutic intervention. SIN3B has been implicated in the initiation of senescence
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, clone DM1A, ascites fluid