Merck
  • First outbreak and subsequent cases of Trypanosoma vivax in the state of Goiás, Brazil.
First outbreak and subsequent cases of Trypanosoma vivax in the state of Goiás, Brazil.
Revista brasileira de parasitologia veterinaria = Brazilian journal of veterinary parasitology : Orgao Oficial do Colegio Brasileiro de Parasitologia Veterinaria (2017-07-06)
Thiago Souza Azeredo Bastos, Adriana Marques Faria, Darling Mélany de Carvalho Madrid, Luciana Cardoso de Bessa, Guido Fontgalland Coelho Linhares, Otavio Luiz Fidelis, Paulo Henrique Sampaio, Breno Cayeiro Cruz, Leonardo Bueno Cruvinel, João Eduardo Nicaretta, Rosangela Zacarias Machado, Alvimar José da Costa, Welber Daniel Zanetti Lopes
ABSTRACT
Trypanosomiasis caused by Trypanosoma vivax has increased the reports in Brazil in the last decade. An outbreak is herein first reported in the state of Goiás, from May 2016 to January 2017. The outbreak start occurred in the city of Ipameri (Goiás) after the introduction of 18 auctioned cows from the state of Minas Gerais. Direct parasitological test (blood smears) and polymerase chain reactions targeting the catL genes diagnosed T. vivax infection. Fifty six cows from a herd of 161 were infected; 12 died during the outbreak and 44 animals persistently positive (by blood smears) even after chemical treatment were discarded. After this first case, five other cases were detected in state of Goiás. The spread of this disease can be linked to the commercialization of animals carrying T. vivax, allied to the iatrogenic transmission practice, using a single needle and syringe for all cows, during oxytocin administration before each milking.
Product Description
Supelco
Diminazene aceturate, analytical standard