Merck
  • Maternal DCAF2 is crucial for maintenance of genome stability during the first cell cycle in mice.
Maternal DCAF2 is crucial for maintenance of genome stability during the first cell cycle in mice.
Journal of cell science (2017-08-19)
Yi-Wen Xu, Lan-Rui Cao, Min Wang, Ying Xu, Xin Wu, Junping Liu, Chao Tong, Heng-Yu Fan
ABSTRACT
Precise regulation of DNA replication and genome integrity is crucial for gametogenesis and early embryogenesis. Cullin ring-finger ubiquitin ligase 4 (CRL4) has multiple functions in the maintenance of germ cell survival, oocyte meiotic maturation, and maternal-zygotic transition in mammals. DDB1-cullin-4-associated factor-2 (DCAF2, also known as DTL or CDT2) is an evolutionarily conserved substrate receptor of CRL4. To determine whether DCAF2 is a key CRL4 substrate adaptor in mammalian oocytes, we generated a novel mouse strain that carries a
Sigma-Aldrich
Mineral oil, light oil, suitable for mouse embryo cell culture, BioXtra
Sigma-Aldrich
M2 medium, With HEPES, without penicillin and streptomycin, liquid, sterile-filtered, suitable for mouse embryo cell culture
Sigma-Aldrich
M16 Medium, With sodium bicarbonate and lactic acid, without penicillin and streptomycin, liquid, sterile-filtered