Merck
  • Phospholipid-drug conjugates as a novel oral drug targeting approach for the treatment of inflammatory bowel disease.
Phospholipid-drug conjugates as a novel oral drug targeting approach for the treatment of inflammatory bowel disease.
European journal of pharmaceutical sciences : official journal of the European Federation for Pharmaceutical Sciences (2017-06-20)
Arik Dahan, Milica Markovic, Svetlana Epstein, Noa Cohen, Ellen M Zimmermann, Aaron Aponick, Shimon Ben-Shabat
ABSTRACT
The enzyme phospholipase A
Sigma-Aldrich
DL-2-Aminocaprylic acid, 99%