Merck
Electron ptychographic microscopy for three-dimensional imaging.
Nature communications (2017-08-02)
Si Gao, Peng Wang, Fucai Zhang, Gerardo T Martinez, Peter D Nellist, Xiaoqing Pan, Angus I Kirkland
ABSTRACT
Knowing the three-dimensional structural information of materials at the nanometer scale is essential to understanding complex material properties. Electron tomography retrieves three-dimensional structural information using a tilt series of two-dimensional images. In this paper, we report an alternative combination of electron ptychography with the inverse multislice method. We demonstrate depth sectioning of a nanostructured material into slices with 0.34 nm lateral resolution and with a corresponding depth resolution of about 24-30 nm. This three-dimensional imaging method has potential applications for the three-dimensional structure determination of a range of objects, ranging from inorganic nanostructures to biological macromolecules.Three-dimensional ptychographic imaging with electrons has remained a challenge because, unlike X-rays, electrons are easily scattered by atoms. Here, Gao et al. extend multi-slice methods to electrons in the multiple scattering regime, paving the way to nanometer-scale 3D structure determination with electrons.
Sigma-Aldrich
Carbon nanotube, multi-walled, >98% carbon basis, O.D. × L 6-13 nm × 2.5-20 μm