Merck
  • Serum based fluorescent assay for evaluating dipeptidyl peptidase I activity in collagen induced arthritis rat model.
Serum based fluorescent assay for evaluating dipeptidyl peptidase I activity in collagen induced arthritis rat model.
Molecular and cellular probes (2016-10-26)
Xiaoqian Liu, Jingjing Wang, Yi Chu, Xiaoying Zhou
ABSTRACT
Dipeptidyl peptidase I (DPPI) is a lysosomal cysteine protease and derived from immune granule cells. It has been suggested playing an important role in the development of rheumatoid arthritis. In this study, a coumarin based fluorescent probe (GF-AFC) was designed and synthesized to evaluate DPPI activity in serum or tissue homogenates of collagen-induced arthritis (CIA) rats, an inflammatory arthropathy model. It was revealed that the fluorescent intensity was significantly increased in a very short time after specific substrate GF-AFC reacted with the DPPI. The fluorophore (AFC) was released to shine after the cleavage reaction which was examined by
Sigma-Aldrich
7-Amino-4-(trifluoromethyl)coumarin, ≥99%
Sigma-Aldrich
Boc-Phe-OH, ≥99.0% (T)