- Archaeal Persisters: Persister Cell Formation as a Stress Response in
Archaeal Persisters: Persister Cell Formation as a Stress Response in
Frontiers in microbiology (2017-09-06)
Julianne Megaw, Brendan F Gilmore
PMID28871247
ABSTRACT
Persister cells are phenotypic variants within a microbial population, which are dormant and transiently tolerant to stress. Persistence has been studied extensively in bacteria, and in eukaryotes to a limited extent, however, it has never been observed in archaea. Using the model haloarchaeon,