Merck
  • A novel animal model of severe pancreatitis in mice and its differences to the rat.
A novel animal model of severe pancreatitis in mice and its differences to the rat.
Surgery (2008-08-19)
Werner Hartwig, Eik Schimmel, Thilo Hackert, Franco Fortunato, Frank Bergmann, Andrea Baczako, Oliver Strobel, Markus W Büchler, Jens Werner
ABSTRACT
A noninvasive model of necrohemorrhagic pancreatitis induced by simultaneous intravenous cerulein/enterokinase (EK) infusion has recently been established in rats. The aim of the present study was to establish this new model in mice and to compare it with the rat model. Male Balb/C mice (20 to 25 g) were used for the experiments. Pancreatitis was induced by simultaneous intravenous infusion of cerulein and EK. Controls were infused with either 0.9% NaCl, cerulein, or EK. Animals were humanely killed 6 hours after start of infusions. Pancreatic and pulmonary injury was assessed by histology, wet-to-dry weight ratio, and myeloperoxidase activity. Systemic cytokine, amylase, and lactate dehydrogenase (LDH) levels in blood were measured to assess pancreatic and systemic inflammatory response. To evaluate the role of protease activity in this model, trypsin, cathepsin B, and elastase activity were measured in pancreatic tissue. Survival experiments were performed to determine survival time and tissue injury in the later course of the disease. Mice with simultaneous cerulein/EK infusion developed marked local and systemic organ injury compared with those animals who received cerulein or EK alone. Pancreatic and pulmonary injury increased with high concentrations of cerulein/EK infusions. Survival decreased in these animals. Whereas acinar cell apoptosis was an early finding, pancreatic necrosis was observed later in the course of the disease. Serum levels of LDH, interleukin (IL)-1 alpha, and IL-1 beta reflected cell damage and the systemic inflammatory response. Protease activity in pancreatic tissue was greatest in animals with simultaneous cerulein/EK infusion. Using intravenous cerulein/EK infusions, a model of lethal acute pancreatitis has been established in mice. Major pancreatic edema, acinar cell apoptosis and necrosis, and pulmonary leukocyte sequestration are characteristic findings in this model. Although pancreatic injury was not as strong as in the rat model, this model may prove useful for future studies in transgenic mice.
Sigma-Aldrich
Enterokinase from porcine intestine, ≥0.5 units/mg solid