Merck
  • The effect of carbon monoxide pretreatment exposure time on the colour stability and quality attributes of vacuum packaged beef steaks.
The effect of carbon monoxide pretreatment exposure time on the colour stability and quality attributes of vacuum packaged beef steaks.
Meat science (2017-03-05)
Lauren Anne Van Rooyen, Paul Allen, Sarah M Crawley, David I O'Connor
ABSTRACT
The effect of 5% CO pretreatments prior to vacuum packaging of beef striploin steaks (Longissimus thoracis et lumborum, LTL) on quality attributes, primarily colour stability was investigated. The aim was to determine the optimum pretreatment that would induce the desirable red colour, while allowing discoloration to occur by the end of a 28-day display period (2°C), so as to not mask spoilage. A range of pretreatment exposure times (1, 3, 5, 7, 9, 15 and 24h) were applied to steaks using a gas mixture of 5% CO, 60% CO
Product Description
Butylhydroxytoluene, European Pharmacopoeia (EP) Reference Standard