- The effect of prefreeze rejuvenation on postthaw storage of red blood cells in AS-3 and SAGM.
The effect of prefreeze rejuvenation on postthaw storage of red blood cells in AS-3 and SAGM.
We investigated whether improving the metabolic status of red blood cell concentrates before freezing could extend the postthaw shelf life beyond 14 days while still meeting the requirements for hemolysis (0.8%) and total adenylate (>82% of original values). At Day 8 after collection, four leukoreduced red blood cell concentrates in saline-adenine-glucose-mannitol (SAGM) were pooled, mixed, and split (n = 4). Of these concentrates, two were rejuvenated in Rejuvesol. In addition, two leukoreduced red blood cell concentrates in phosphate-adenine-glucose-guanosine-gluconate-mannitol (PAGGGM) were pooled, mixed, and split at Day 8 after collection (n = 4). All concentrates were glycerolized, frozen, and stored for at least 2 weeks at -80°C. After thawing and deglycerolization, from each pair, one red blood cell concentrate was resuspended in SAGM, and one was suspended in AS-3. During postthaw storage at 2 to 6°C for 35 days, all concentrates were sampled weekly and analyzed for hematologic, metabolic, and morphologic parameters. Both Rejuvesol and PAGGGM treatment produced increased adenosine triphosphate and total adenylate and 2,3-diphosphoglycerate levels compared with untreated red blood cell concentrates. Regardless of prefreeze Rejuvesol or PAGGGM treatment, postthaw hemolysis remained below 0.8% during 7 days in SAGM and during 35 days in AS-3. At Day 35 of postthaw storage in AS-3, total adenylate in nonrejuvenated red blood cell concentrates had decreased to 72% of the original values; whereas, in prefreeze Rejuvesol-treated and PAGGGM-treated concentrates, adenylate values were still were at 101% and 98%, respectively. Based on maximum allowable hemolysis of 0.8% and total adenylate content greater than 82% of the original value, thawed, prefreeze Rejuvesol-treated or PAGGGM-treated red blood cell concentrates can be stored for 35 days at 2 to 6ºC in AS-3.