Merck
  • Toxicity assessment of zero valent iron nanoparticles on Artemia salina.
Toxicity assessment of zero valent iron nanoparticles on Artemia salina.
Environmental toxicology (2017-01-20)
Deepak Kumar, Rajdeep Roy, Abhinav Parashar, Ashok M Raichur, Natarajan Chandrasekaran, Anita Mukherjee, Amitava Mukherjee
ABSTRACT
The present study deals with the toxicity assessment of two differently synthesized zero valent iron nanoparticles (nZVI, chemical and biological) as well as Fe
Sigma-Aldrich
Glutathione Reductase Assay Kit, Sufficient for 100 colorimetric tests