Merck
  • N-arachidonoyl-serotonin, a dual FAAH and TRPV1 blocker, inhibits the retrieval of contextual fear memory: Role of the cannabinoid CB1 receptor in the dorsal hippocampus.
N-arachidonoyl-serotonin, a dual FAAH and TRPV1 blocker, inhibits the retrieval of contextual fear memory: Role of the cannabinoid CB1 receptor in the dorsal hippocampus.
Journal of psychopharmacology (Oxford, England) (2017-06-07)
Pedro H Gobira, Isabel V Lima, Luara A Batista, Antônio C de Oliveira, Leonardo B Resstel, Carsten T Wotjak, Daniele C Aguiar, Fabricio A Moreira
ABSTRACT
Anandamide, an endocannabinoid, inhibits aversive responses by activating the CB
Sigma-Aldrich
URB597, ≥98% (HPLC), powder
Sigma-Aldrich
AM251, >98% (HPLC), solid