Merck
  • The telomere-binding protein TRF2 is required for metronomic therapeutic effects of gemcitabine and capecitabine.
The telomere-binding protein TRF2 is required for metronomic therapeutic effects of gemcitabine and capecitabine.
Biochimica et biophysica acta (2017-01-16)
Wei-Ping Lee, Keng-Hsin Lan, Chung-Pin Li, Yee Chao, Ming-Chih Hou, Han-Chieh Lin, Shou-Dong Lee
ABSTRACT
Gemcitabine and capecitabine are two effective anticancer agents against solid tumors. The pharmacological mechanisms have been known as incorporation into DNA and thereby inhibition of DNA synthesis. When used as metronomic chemotherapy, they may inhibit angiogenesis and induce immunity. In our previous study, we showed that low-dose gemcitabine caused telomere shortening by stabilizing TRF2 that was required for XPF-dependent telomere loss. In this report, we established a SKOV3.ip1 ascites cell model. Tumor-bearing mice were treated with low-dose gemcitabine (GEM) or capecitabine (CAP). Both GEM and CAP caused telomere shortening and increased expression of TRF2 with improved ascites in nude mice and decreased in vitro clonogenic activity. TRF2 knockdown altered telomeres to a shortened but new status that may evade XPF-dependent telomere loss and conferred resistance of SKOV3.ip1 ascites cells to low-dose GEM and CAP. Our study provides a new mechanism of metronomic chemotherapy i.e. TRF2 is required for metronomic therapeutic effects of gemcitabine and capecitabine.
Sigma-Aldrich
MISSION® esiRNA, targeting human ERCC1