Merck
  • Sensitive Versatile Fluorogenic Transmembrane Peptide Substrates for Rhomboid Intramembrane Proteases.
Sensitive Versatile Fluorogenic Transmembrane Peptide Substrates for Rhomboid Intramembrane Proteases.
The Journal of biological chemistry (2017-01-11)
Anežka Tichá, Stancho Stanchev, Jan Škerle, Jakub Began, Marek Ingr, Kateřina Švehlová, Lucie Polovinkin, Martin Růžička, Lucie Bednárová, Romana Hadravová, Edita Poláchová, Petra Rampírová, Jana Březinová, Václav Kašička, Pavel Majer, Kvido Strisovsky
ABSTRACT
Rhomboid proteases are increasingly being explored as potential drug targets, but their potent and specific inhibitors are not available, and strategies for inhibitor development are hampered by the lack of widely usable and easily modifiable
Sigma-Aldrich
3-(Dimethylamino)phenol, 97%