Merck
Conjugated polymer with carboxylate groups-Hg
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2017-01-17)
Hongyu Mi, Mingming Guan, Jilin Liu, Hongyan Shan, Qiang Fei, Yanfu Huan, Guodong Feng
ABSTRACT
In this work, a turn on fluorescent sensor, based on Hg
Sigma-Aldrich
1,4-Diethynylbenzene, 96%
Sigma-Aldrich
Tetrakis(triphenylphosphine)palladium(0), 99%