Merck
Dissociative recombination of HCl
The Journal of chemical physics (2017-01-05)
Justin P Wiens, Thomas M Miller, Nicholas S Shuman, Albert A Viggiano
ABSTRACT
Dissociative recombination of electrons with HCl
Sigma-Aldrich
Argon, Messer® CANGas, 99.999%