Merck
  • Nestin expression in end-stage disease in dystrophin-deficient heart: implications for regeneration from endogenous cardiac stem cells.
Nestin expression in end-stage disease in dystrophin-deficient heart: implications for regeneration from endogenous cardiac stem cells.
Stem cells translational medicine (2013-09-27)
Suzanne E Berry, Peter Andruszkiewicz, Ju Lan Chun, Jun Hong
ABSTRACT
Nestin(+) cardiac stem cells differentiate into striated cells following myocardial infarct. Transplantation of exogenous stem cells into myocardium of a murine model for Duchenne muscular dystrophy (DMD) increased proliferation of endogenous nestin(+) stem cells and resulted in the appearance of nestin(+) striated cells. This correlated with, and may be responsible for, prevention of dilated cardiomyopathy. We examined nestin(+) stem cells in the myocardium of dystrophin/utrophin-deficient (mdx/utrn(-/-)) mice, a model for DMD. We found that 92% of nestin(+) interstitial cells expressed Flk-1, a marker present on cardiac progenitor cells that differentiate into the cardiac lineage, and that a subset expressed Sca-1, present on adult cardiac cells that become cardiomyocytes. Nestin(+) interstitial cells maintained expression of Flk-1 but lost Sca-1 expression with age and were present in lower numbers in dystrophin-deficient heart than in wild-type heart. Unexpectedly, large clusters of nestin(+) striated cells ranging in size from 20 to 250 cells and extending up to 500 μm were present in mdx/utrn(-/-) heart near the end stage of disease. These cells were also present in dystrophin-deficient mdx/utrn(+/-) and mdx heart but not wild-type heart. Nestin(+) striated cells expressed cardiac troponin I, desmin, and Connexin 43 and correlated with proinflammatory CD68(+) macrophages. Elongated nestin(+) interstitial cells with striations were observed that did not express Flk-1 or the late cardiac marker cardiac troponin I but strongly expressed the early cardiac marker desmin. Nestin was also detected in endothelial and smooth muscle cells. These data indicate that new cardiomyocytes form in dystrophic heart, and nestin(+) interstitial cells may generate them in addition to other cells of the cardiac lineage.
Sigma-Aldrich
Anti-Troponin I Antibody, a.a. 186-192, clone C5, clone C5, Chemicon®, from mouse
Sigma-Aldrich
Anti-Connexin-43 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution