Merck
  • The induction of apoptosis in SGC-7901 cells through the ROS-mediated mitochondrial dysfunction pathway by a Ir(III) complex.
The induction of apoptosis in SGC-7901 cells through the ROS-mediated mitochondrial dysfunction pathway by a Ir(III) complex.
Journal of biological inorganic chemistry : JBIC : a publication of the Society of Biological Inorganic Chemistry (2016-11-01)
Cheng Zhang, Shang-Hai Lai, Chuan-Chuan Zeng, Bing Tang, Dan Wan, De-Gang Xing, Yun-Jun Liu
ABSTRACT
A new ligand BTCP and its iridium(III) complex [Ir(ppy)
Sigma-Aldrich
Bicyclo[2.2.1]hept-5-ene-2-carboxaldehyde, 95%